n-Butyllithium, 2.5M solution in hexanes, AcroSeal™
n-Butyllithium, 2.5M solution in hexanes, AcroSeal™
n-Butyllithium, 2.5M solution in hexanes, AcroSeal™
n-Butyllithium, 2.5M solution in hexanes, AcroSeal™
Thermo Scientific Chemicals

n-Butyllithium, 2.5M solution in hexanes, AcroSeal™

CAS: 109-72-8 | C4H9Li | 64.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
100 mL
800 mL
Packaging:
View all
Glass bottle
Catalog number 213350500
Price (EUR)
44,65
Online Exclusive
49,80
Save 5,15 (10%)
Each
Quantity:
50 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
44,65
Online Exclusive
49,80
Save 5,15 (10%)
Each
Chemical Identifiers
CAS109-72-8
IUPAC Namebutyllithium
Molecular FormulaC4H9Li
InChI KeyMZRVEZGGRBJDDB-UHFFFAOYSA-N
SMILES[Li]CCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear to hazy colorless to yellow
Appearance (Form)Solution
AppearanceMay develop some turbidity or precipitate
Titration Active base22.0 to 24.0 %
Titration Active base2.37 to 2.59 M
View more

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

RUO – Research Use Only