Glycyrrhizic acid monoammonium salt, 98+%
Glycyrrhizic acid monoammonium salt, 98+%
Glycyrrhizic acid monoammonium salt, 98+%
Thermo Scientific Chemicals

Glycyrrhizic acid monoammonium salt, 98+%

CAS: 53956-04-0 | C42H65NO16 | 839.97 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 213750250
Price (EUR)
80,65
Online Exclusive
89,60
Save 8,95 (10%)
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
80,65
Online Exclusive
89,60
Save 8,95 (10%)
Each
Chemical Identifiers
CAS53956-04-0
IUPAC Name(2S,3S,4S,5R,6S)-6-{[(3S,4aR,6aR,6bS,8aS,11S,12aR,14aR,14bS)-11-carboxy-4,4,6a,6b,8a,11,14b-heptamethyl-14-oxo-1,2,3,4,4a,5,6,6a,6b,7,8,8a,9,10,11,12,12a,14,14a,14b-icosahydropicen-3-yl]oxy}-5-{[(2R,3R,4S,5S,6S)-6-carboxy-3,4,5-trihydroxyoxan-2-yl]oxy}-3,4-dihydroxyoxane-2-carboxylic acid amine
Molecular FormulaC42H65NO16
InChI KeyILRKKHJEINIICQ-OOFFSTKBSA-N
SMILESN.CC1(C)[C@H](CC[C@@]2(C)[C@H]1CC[C@]1(C)[C@@H]2C(=O)C=C2[C@@H]3C[C@](C)(CC[C@]3(C)CC[C@@]12C)C(O)=O)O[C@H]1O[C@@H]([C@@H](O)[C@H](O)[C@H]1O[C@@H]1O[C@@H]([C@@H](O)[C@H](O)[C@H]1O)C(O)=O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Heavy metals=<20 ppm
Appearance (Color)White to light yellow
Infrared spectrumConforms
Water=<6.0 % (K.F.)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only