Squalane, 99%, Thermo Scientific Chemicals
Squalane, 99%, Thermo Scientific Chemicals
Squalane, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Squalane, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
2.5 L
Packaging:
Glass bottle
Catalog number 215355000
Price (EUR)
-
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS111-01-3
IUPAC Name2,6,10,15,19,23-hexamethyltetracosane
Molecular FormulaC30H62
InChI KeyPRAKJMSDJKAYCZ-UHFFFAOYNA-N
SMILESCC(C)CCCC(C)CCCC(C)CCCCC(C)CCCC(C)CCCC(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Infrared spectrumConforms
Refractive index1.4510 to 1.4530 (20°C, 589 nm)
Iodine value=<1 g I/100 g
Appearance (Form)Liquid
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only