Boric acid, 99.5%, for analysis
Boric acid, 99.5%, for analysis
Boric acid, 99.5%, for analysis
Thermo Scientific Chemicals

Boric acid, 99.5%, for analysis

CAS: 10043-35-3 | BH3O3 | 61.83 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2.5 kg
10 kg
Packaging:
View all
Plastic bottle
Catalog number 217080025
Price (EUR)
163,00
Each
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (EUR)
163,00
Each
Chemical Identifiers
CAS10043-35-3
IUPAC Nameboric acid
Molecular FormulaBH3O3
InChI KeyKGBXLFKZBHKPEV-UHFFFAOYSA-N
SMILESOB(O)O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Titration with NaOH>=99.4 %
Phosphate (PO4)=<5 ppm
Magnesium (Mg)=<5 ppm
Arsenic (As)=<0.5 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer, Lewis acid
RUO – Research Use Only