Cerium(III) nitrate hexahydrate, 99.5%
Cerium(III) nitrate hexahydrate, 99.5%
Cerium(III) nitrate hexahydrate, 99.5%
Thermo Scientific Chemicals

Cerium(III) nitrate hexahydrate, 99.5%

CAS: 10294-41-4 | CeH12N3O15 | 434.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number 218690025
Price (EUR)
682,65
Online exclusive
758,00
Save 75,35 (10%)
Each
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (EUR)
682,65
Online exclusive
758,00
Save 75,35 (10%)
Each
Chemical Identifiers
CAS10294-41-4
IUPAC Namecerium(3+) hexahydrate trinitrate
Molecular FormulaCeH12N3O15
InChI KeyQQZMWMKOWKGPQY-UHFFFAOYSA-N
SMILESO.O.O.O.O.O.[Ce+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to white
Appearance (Form)Powder or crystals and/or chunks
Assay>=99.4 % (CeO2/TREO basis)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only