Chromium(III) nitrate nonahydrate, 99%
Chromium(III) nitrate nonahydrate, 99%
Chromium(III) nitrate nonahydrate, 99%
Thermo Scientific Chemicals

Chromium(III) nitrate nonahydrate, 99%

CAS: 7789-02-8 | CrN3O9·9H2O | 400.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
1 kg
5 kg
Packaging:
Plastic bottle
Catalog number 219205000
Price (EUR)
49,65
Online exclusive
55,10
Save 5,45 (10%)
Each
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (EUR)
49,65
Online exclusive
55,10
Save 5,45 (10%)
Each
Chemical Identifiers
CAS7789-02-8
IUPAC Namechromium(3+) trinitrate
Molecular FormulaCrN3O9
InChI KeyPHFQLYPOURZARY-UHFFFAOYSA-N
SMILES[Cr+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Calcium (Ca)=<50 ppm
Copper (Cu)=<10 ppm
Magnesium (Mg)=<50 ppm
Sodium (Na)=<50 ppm
Appearance (Form)Crystalline chunks
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only