2-Ethylimidazole, 99%
2-Ethylimidazole, 99%
2-Ethylimidazole, 99%
Thermo Scientific Chemicals

2-Ethylimidazole, 99%

CAS: 1072-62-4 | C5H8N2 | 96.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
View all
Plastic Bottle
Catalog number 219280025
Price (EUR)
419,00
Each
Quantity:
2.5 kg
Packaging:
Plastic Bottle
Request bulk or custom format
Price (EUR)
419,00
Each
Chemical Identifiers
CAS1072-62-4
IUPAC Name2-ethyl-1H-imidazole
Molecular FormulaC5H8N2
InChI KeyPQAMFDRRWURCFQ-UHFFFAOYSA-N
SMILESCCC1=NC=CN1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Flakes
GC>=98.5 %
Melting point83°C to 87°C
GC=<1.0 % Imidazole
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only