sec-Butanol, 99+%, for analysis
sec-Butanol, 99+%, for analysis
sec-Butanol, 99+%, for analysis
Thermo Scientific Chemicals

sec-Butanol, 99+%, for analysis

CAS: 78-92-2 | C4H10O | 74.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
Catalog number 220290010
Price (EUR)
64,30
Each
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (EUR)
64,30
Each
Chemical Identifiers
CAS78-92-2
IUPAC Namebutan-2-ol
Molecular FormulaC4H10O
InChI KeyBTANRVKWQNVYAZ-UHFFFAOYNA-N
SMILESCCC(C)O
View more
SpecificationsSpecification SheetSpecification Sheet
Color scale=<10 APHA
Water=<0.2 %
Iron (Fe)=<0.5 ppm
Free acid (C4H8O2)=<0.003 %
Appearance (Form)Clear liquid
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only