Phenyl vinyl sulfone, 98%
Phenyl vinyl sulfone, 98%
Phenyl vinyl sulfone, 98%
Thermo Scientific Chemicals

Phenyl vinyl sulfone, 98%

CAS: 5535-48-8 | C8H8O2S | 168.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number 221250010
Price (EUR)
30,00
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
30,00
Each
Chemical Identifiers
CAS5535-48-8
IUPAC Name(ethenesulfonyl)benzene
Molecular FormulaC8H8O2S
InChI KeyUJTPZISIAWDGFF-UHFFFAOYSA-N
SMILESC=CS(=O)(=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to beige
GC>=97.5 %
Infrared spectrumConforms
Appearance (Form)Powder and/or chunks
Melting point66°C to 70°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only