N-Methyl-N-(trimethylsilyl)trifluoroacetamide, 97%
N-Methyl-N-(trimethylsilyl)trifluoroacetamide, 97%
N-Methyl-N-(trimethylsilyl)trifluoroacetamide, 97%
Thermo Scientific Chemicals

N-Methyl-N-(trimethylsilyl)trifluoroacetamide, 97%

CAS: 24589-78-4 | C6H12F3NOSi | 199.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
View all
Glass bottle
Catalog number 221581000
Price (EUR)
1.013,65
線上優惠
1.126,00
Save 112,35 (10%)
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
1.013,65
線上優惠
1.126,00
Save 112,35 (10%)
Each
Chemical Identifiers
CAS24589-78-4
IUPAC Name2,2,2-trifluoro-N-methyl-N-(trimethylsilyl)acetamide
Molecular FormulaC6H12F3NOSi
InChI KeyMSPCIZMDDUQPGJ-UHFFFAOYSA-N
SMILESCN(C(=O)C(F)(F)F)[Si](C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Appearance (Color)Clear colorless to light yellow
GC>=96.0 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Alkylation reagent, Silylation reagent
RUO – Research Use Only