Bromoacetonitrile, 97%
Bromoacetonitrile, 97%
Bromoacetonitrile, 97%
Thermo Scientific Chemicals

Bromoacetonitrile, 97%

CAS: 590-17-0 | C2H2BrN | 119.95 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 221860050
Price (EUR)
37,70
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
37,70
Each
Chemical Identifiers
CAS590-17-0
IUPAC Name2-bromoacetonitrile
Molecular FormulaC2H2BrN
InChI KeyREXUYBKPWIPONM-UHFFFAOYSA-N
SMILESBrCC#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Infrared spectrumConforms
Refractive index1.4780 to 1.4810 (20°C, 589 nm)
Appearance (Form)Liquid
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only