1-Bromo-2-butanone, 90%, stabilized, Thermo Scientific Chemicals
1-Bromo-2-butanone, 90%, stabilized, Thermo Scientific Chemicals
1-Bromo-2-butanone, 90%, stabilized, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Bromo-2-butanone, 90%, stabilized, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 222450250
Price (EUR)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS816-40-0
IUPAC Name1-bromobutan-2-one
Molecular FormulaC4H7BrO
InChI KeyCCXQVBSQUQCEEO-UHFFFAOYSA-N
SMILESCCC(=O)CBr
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=88 %
Refractive index1.4650 to 1.4690 (20°C, 589 nm)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only