L(+)-Potassium hydrogen tartrate, 99%
L(+)-Potassium hydrogen tartrate, 99%
L(+)-Potassium hydrogen tartrate, 99%
Thermo Scientific Chemicals

L(+)-Potassium hydrogen tartrate, 99%

CAS: 868-14-4 | C4H5KO6 | 188.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
5 kg
Packaging:
View all
Plastic drum
Catalog number 222670051
Price (EUR)
164,65
Online exclusive
183,00
Save 18,35 (10%)
Each
Quantity:
5 kg
Packaging:
Plastic drum
Request bulk or custom format
Price (EUR)
164,65
Online exclusive
183,00
Save 18,35 (10%)
Each
Chemical Identifiers
CAS868-14-4
IUPAC Namepotassium (2R,3R)-3-carboxy-2,3-dihydroxypropanoate
Molecular FormulaC4H5KO6
InChI KeyKYKNRZGSIGMXFH-ZVGUSBNCSA-M
SMILES[K+].O[C@H]([C@@H](O)C([O-])=O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to white
Loss on drying=<0.5 % (105°C)
Specific optical rotationon dried substance
Sulfate (SO4)=<500 ppm
Appearance (Form)Powder or crystals
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only