Sodium hypophosphite monohydrate, for analysis
Sodium hypophosphite monohydrate, for analysis
Sodium hypophosphite monohydrate, for analysis
Sodium hypophosphite monohydrate, for analysis
Thermo Scientific Chemicals

Sodium hypophosphite monohydrate, for analysis

CAS: 10039-56-2 | H4NaO3P | 105.99 g/mol
Have Questions?
Catalog NumberQuantityPackaging
222791000100 gPlastic bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 222791000
Price (EUR)
-
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS10039-56-2
IUPAC Namesodium hydrate phosphinate
Molecular FormulaH4NaO3P
InChI KeyPLZNPHDJGFDNRM-UHFFFAOYSA-M
SMILESO.[Na+].[O-][PH2]=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Titration99.0 to 103.0 %
Arsenic (As)=<2 ppm
Iron (Fe)=<5 ppm
Lead (Pb)=<5 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only