Phenylselenyl bromide, 98%
Phenylselenyl bromide, 98%
Phenylselenyl bromide, 98%
Thermo Scientific Chemicals

Phenylselenyl bromide, 98%

CAS: 34837-55-3 | C6H5BrSe | 235.97 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 223060050
Price (EUR)
72,50
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
72,50
Each
Chemical Identifiers
CAS34837-55-3
IUPAC Name(phenylselanyl)bromane
Molecular FormulaC6H5BrSe
InChI KeyLCEFEIBEOBPPSJ-UHFFFAOYSA-N
SMILESBr[Se]C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Titration Argentometric>=97.5 %
Appearance (Color)Dark brown to black
Infrared spectrumConforms
Appearance (Form)Crystals
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only