Cadmium oxide, 99%, pure
Cadmium oxide, 99%, pure
Cadmium oxide, 99%, pure
Thermo Scientific Chemicals

Cadmium oxide, 99%, pure

CAS: 1306-19-0 | CdO | 128.41 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 223792500
Price (EUR)
40,70
Each
Quantity:
250 g
Request bulk or custom format
Price (EUR)
40,70
Each
Chemical Identifiers
CAS1306-19-0
IUPAC Namecadmium(2+) oxidandiide
Molecular FormulaCdO
InChI KeyCFEAAQFZALKQPA-UHFFFAOYSA-N
SMILES[O--].[Cd++]
View more
SpecificationsSpecification SheetSpecification Sheet
Titration Complexometric>=98.5 %
Appearance (Form)Powder
Appearance (Color)Red-brown to brown
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only