Bis(1,5-cyclooctadiene)nickel(0)
Bis(1,5-cyclooctadiene)nickel(0)
Bis(1,5-cyclooctadiene)nickel(0)
Thermo Scientific Chemicals

Bis(1,5-cyclooctadiene)nickel(0)

CAS: 1295-35-8 | C16H24Ni | 275.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number 223970010
Price (EUR)
114,00
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
114,00
Each
Chemical Identifiers
CAS1295-35-8
IUPAC Namebis((1Z,5Z)-cycloocta-1,5-diene) nickel
Molecular FormulaC16H24Ni
InChI KeyJRTIUDXYIUKIIE-KZUMESAESA-N
SMILES[Ni].C1C\C=C/CC\C=C/1.C1C\C=C/CC\C=C/1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to orange
Nickel (Ni)20.0 to 22.6 % (with dimethylglyoxime)
Appearance (Form)Powder or crystals
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only