L(-)-Sorbose, 98%
L(-)-Sorbose, 98%
L(-)-Sorbose, 98%
Thermo Scientific Chemicals

L(-)-Sorbose, 98%

CAS: 87-79-6 | C6H12O6 | 180.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
250 g
1 kg
Packaging:
Plastic bottle
Catalog number 225962500
Price (EUR)
421,00
Each
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (EUR)
421,00
Each
Chemical Identifiers
CAS87-79-6
IUPAC Name1,3,4,5,6-pentahydroxyhexan-2-one
Molecular FormulaC6H12O6
InChI KeyBJHIKXHVCXFQLS-UHFFFAOYNA-N
SMILESOCC(O)C(O)C(O)C(=O)CO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals and/or chunks
Infrared spectrumConforms
HPLC>=97.5 %
Loss on drying=<1 %
Specific optical rotation-41° to -45° (20°C, 589 nm) (c=5, H2O)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only