N-Acetyl-DL-tryptophan, 98+%, extra pure
N-Acetyl-DL-tryptophan, 98+%, extra pure
N-Acetyl-DL-tryptophan, 98+%, extra pure
Thermo Scientific Chemicals

N-Acetyl-DL-tryptophan, 98+%, extra pure

CAS: 87-32-1 | C13H14N2O3 | 246.27 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 227151000
Price (EUR)
75,65
Online exclusive
83,50
Save 7,85 (9%)
Each
Quantity:
100 g
Request bulk or custom format
Price (EUR)
75,65
Online exclusive
83,50
Save 7,85 (9%)
Each
Chemical Identifiers
CAS87-32-1
IUPAC Name2-acetamido-3-(1H-indol-3-yl)propanoic acid
Molecular FormulaC13H14N2O3
InChI KeyDZTHIGRZJZPRDV-UHFFFAOYSA-N
SMILESCC(=O)NC(CC1=CNC2=CC=CC=C12)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Loss on drying=<0.5 % (105°C)
Sulfated ash=<0.2 %
Appearance (Color)White to beige
Infrared spectrumConforms
Titration excess NaOH98.0 to 101.0 % (On dry substance)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only