L-Aspartyl-L-phenylalanine methyl ester, 98%
L-Aspartyl-L-phenylalanine methyl ester, 98%
L-Aspartyl-L-phenylalanine methyl ester, 98%
Thermo Scientific Chemicals

L-Aspartyl-L-phenylalanine methyl ester, 98%

CAS: 22839-47-0 | C14H18N2O5 | 294.31 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Packaging:
Glass Bottle
Catalog number 228650250
Price (EUR)
852,00
Each
Quantity:
25 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (EUR)
852,00
Each
Chemical Identifiers
CAS22839-47-0
IUPAC Name(3S)-3-amino-3-{[(2S)-1-methoxy-1-oxo-3-phenylpropan-2-yl]carbamoyl}propanoic acid
Molecular FormulaC14H18N2O5
InChI KeyIAOZJIPTCAWIRG-QWRGUYRKSA-N
SMILESCOC(=O)[C@H](CC1=CC=CC=C1)NC(=O)[C@@H](N)CC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Heavy metals (as Pb)=<10 ppm
UV transmittance>=95 % (c=1, 2 N HCl, 430 nm) 1 cm cell
Specific optical rotation+12.5° to +17.5° (20°C, 589 nm) (c=4, 15 N formic acid)
Specific optical rotationwithin 30 min
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only