8-Azaguanine 98%
8-Azaguanine 98%
8-Azaguanine 98%
Thermo Scientific Chemicals

8-Azaguanine 98%

CAS: 134-58-7 | C4H4N6O | 152.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 229920010
Price (EUR)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS134-58-7
IUPAC Name5-amino-2H,3H,7H-[1,2,3]triazolo[4,5-d]pyrimidin-7-one
Molecular FormulaC4H4N6O
InChI KeyLPXQRXLUHJKZIE-UHFFFAOYSA-N
SMILESNC1=NC(=O)C2=NNNC2=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
HPLC>=94 %
Loss on drying=<5 %
Appearance (Color)White to yellow to light orange
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only