2-Thiopheneacetyl chloride, 99%
2-Thiopheneacetyl chloride, 99%
2-Thiopheneacetyl chloride, 99%
Thermo Scientific Chemicals

2-Thiopheneacetyl chloride, 99%

CAS: 39098-97-0 | C6H5ClOS | 160.62 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
50 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 232230500
Price (EUR)
137,00
Each
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
137,00
Each
Chemical Identifiers
CAS39098-97-0
IUPAC Name2-(thiophen-2-yl)acetyl chloride
Molecular FormulaC6H5ClOS
InChI KeyAJYXPNIENRLELY-UHFFFAOYSA-N
SMILESClC(=O)CC1=CC=CS1
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=98.5 %
Free acid=<2 %
Appearance (Color)Clear brown
Infrared spectrumConforms
Refractive index1.5500 to 1.5520 (20°C, 589 nm)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only