3-Hydroxypicolinic acid, 98%
3-Hydroxypicolinic acid, 98%
3-Hydroxypicolinic acid, 98%
3-Hydroxypicolinic acid, 98%
Thermo Scientific Chemicals

3-Hydroxypicolinic acid, 98%

CAS: 874-24-8 | C6H4NO3 | 138.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 241050250
Price (EUR)
386,00
Each
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
386,00
Each
Add to cart
Chemical Identifiers
CAS874-24-8
IUPAC Name3-hydroxypyridine-2-carboxylate
Molecular FormulaC6H4NO3
InChI KeyBRARRAHGNDUELT-UHFFFAOYSA-M
SMILESOC1=CC=CN=C1C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
HPLC>=97.5 %
Appearance (Form)Powder
Melting point208°C to 214°C
Appearance (Color)Beige to brown
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only