3-Thiophenecarboxylic acid, 99%
3-Thiophenecarboxylic acid, 99%
3-Thiophenecarboxylic acid, 99%
Thermo Scientific Chemicals

3-Thiophenecarboxylic acid, 99%

CAS: 88-13-1 | C5H4O2S | 128.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 243110050
Price (EUR)
72,40
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
72,40
Each
Chemical Identifiers
CAS88-13-1
IUPAC Namethiophene-3-carboxylic acid
Molecular FormulaC5H4O2S
InChI KeyYNVOMSDITJMNET-UHFFFAOYSA-N
SMILESOC(=O)C1=CSC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Appearance (Color)White to light yellow
Infrared spectrumConforms
Titration with NaOH>=98.5 %
Melting point137°C to 141°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only