2-Hydroxypyridine-N-oxide, 97%
2-Hydroxypyridine-N-oxide, 97%
2-Hydroxypyridine-N-oxide, 97%
Thermo Scientific Chemicals

2-Hydroxypyridine-N-oxide, 97%

CAS: 13161-30-3 | C5H5NO2 | 111.1 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number 243795000
Price (EUR)
488,00
Each
Quantity:
500 g
Request bulk or custom format
Price (EUR)
488,00
Each
Chemical Identifiers
CAS13161-30-3
IUPAC Name1-hydroxy-1,2-dihydropyridin-2-one
Molecular FormulaC5H5NO2
InChI KeySNUSZUYTMHKCPM-UHFFFAOYSA-N
SMILESON1C=CC=CC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Off-white to beige
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point147°C to 152°C
Titration with NaOH>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only