2,2,3,3,4,4,4-Heptafluoro-1-butanol, 95%
2,2,3,3,4,4,4-Heptafluoro-1-butanol, 95%
2,2,3,3,4,4,4-Heptafluoro-1-butanol, 95%
2,2,3,3,4,4,4-Heptafluoro-1-butanol, 95%
Thermo Scientific Chemicals

2,2,3,3,4,4,4-Heptafluoro-1-butanol, 95%

CAS: 375-01-9 | C4H3F7O | 200.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Packaging:
Glass bottle
Catalog number 243890500
Price (EUR)
217,00
Each
Add to cart
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
217,00
Each
Add to cart
Chemical Identifiers
CAS375-01-9
IUPAC Name2,2,3,3,4,4,4-heptafluorobutan-1-ol
Molecular FormulaC4H3F7O
InChI KeyWXJFKAZDSQLPBX-UHFFFAOYSA-N
SMILESOCC(F)(F)C(F)(F)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=94 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only