Propyl chloroformate, 98%
Propyl chloroformate, 98%
Propyl chloroformate, 98%
Thermo Scientific™

Propyl chloroformate, 98%

CAS: 109-61-5 | C4H7ClO2 | 122.55 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 245290250
Price (EUR)
64,65
Online exclusive
71,80
Save 7,15 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
64,65
Online exclusive
71,80
Save 7,15 (10%)
Each
Chemical Identifiers
CAS109-61-5
IUPAC Namepropyl carbonochloridate
Molecular FormulaC4H7ClO2
InChI KeyQQKDTTWZXHEGAQ-UHFFFAOYSA-N
SMILESCCCOC(Cl)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
GC>=97.5 %
Refractive index1.4030 to 1.4050 (20°C, 589 nm)
Appearance (Form)Liquid
Appearance (Color)Clear colorless to yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only