2-Methyl-5-isopropylaniline, Thermo Scientific Chemicals
2-Methyl-5-isopropylaniline, Thermo Scientific Chemicals
2-Methyl-5-isopropylaniline, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Methyl-5-isopropylaniline, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number 248790050
Price (EUR)
324,00
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
324,00
Each
Chemical Identifiers
CAS2051-53-8
IUPAC Name2-methyl-5-(propan-2-yl)aniline
Molecular FormulaC10H15N
InChI KeyYKWALWNGEXPARQ-UHFFFAOYSA-N
SMILESCC(C)C1=CC=C(C)C(N)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.5350 to 1.5450 (20°C, 589 nm)
Appearance (Color)Clear colorless to yellow to brown
AppearanceLiquid
Infrared spectrumConforms
Water=<2.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only