Product Image
Thermo Scientific Chemicals

Ethoxyquin

CAS: 91-53-2 | C14H19NO | 217.31 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 255630250
Price (EUR)
31,90
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (EUR)
31,90
Each
Add to cart
Chemical Identifiers
CAS91-53-2
IUPAC Name6-ethoxy-2,2,4-trimethyl-1,2-dihydroquinoline
Molecular FormulaC14H19NO
InChI KeyDECIPOUIJURFOJ-UHFFFAOYSA-N
SMILESCCOC1=CC=C2NC(C)(C)C=C(C)C2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.5720 to 1.5750 (20°C, 589 nm)
Appearance (Color)Yelow to red to black
Infrared spectrumConforms
GC>=88 %
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only