D(-)-Tagatose, 99+%
D(-)-Tagatose, 99+%
D(-)-Tagatose, 99+%
Thermo Scientific Chemicals

D(-)-Tagatose, 99+%

D(-)-Tagatose, >-99.0%, C6H12O6, CAS Number-87-81-0, tagatose, d, d--tagatose, d-lyxo-hex-2-ulose, 3s,4s,5r-1,3,4,5,6-pentahydroxyhexan-2-one, unii-t7a20y888y, naturlose, tagatose nf, lyxo-2-hexulose, keto-d-tagatose, 1g, (5% soln. in H2O) clear colorless to almost colorless, 99+%, 01, 4414 | CAS: 87-81-0 | C6H12O6 | 180.156 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
Catalog number 257890050
Price (EUR)
416,00
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
416,00
Each
Chemical Identifiers
CAS87-81-0
IUPAC Name(3S,4S,5R)-2-(hydroxymethyl)oxane-2,3,4,5-tetrol
Molecular FormulaC6H12O6
InChI KeyLKDRXBCSQODPBY-OEXCPVAWSA-N
SMILESOCC1(O)OC[C@@H](O)[C@H](O)[C@@H]1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Appearance (Form)Crystalline powder
HPLC>=99.0 %
Infrared spectrumConforms
Loss on drying=<1.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only