1-Ethoxy-2-propanol, 95%
1-Ethoxy-2-propanol, 95%
1-Ethoxy-2-propanol, 95%
Thermo Scientific Chemicals

1-Ethoxy-2-propanol, 95%

CAS: 1569-02-4 | C5H12O2 | 104.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
10 L
Packaging:
View all
Glass Bottle
Catalog number 259050010
Price (EUR)
53,65
온라인 행사
59,60
Save 5,95
Each
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (EUR)
53,65
온라인 행사
59,60
Save 5,95
Each
Chemical Identifiers
CAS1569-02-4
IUPAC Name1-ethoxypropan-2-ol
Molecular FormulaC5H12O2
InChI KeyJOLQKTGDSGKSKJ-UHFFFAOYNA-N
SMILESCCOCC(C)O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=94 %
GC=<6 % (2-Ethoxy-1-propanol)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only