Butyric acid sodium salt, 98%
Butyric acid sodium salt, 98%
Butyric acid sodium salt, 98%
Thermo Scientific Chemicals

Butyric acid sodium salt, 98%

CAS: 156-54-7 | C4H7NaO2 | 110.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
500 g
Packaging:
View all
Glass Bottle
Catalog number 263195000
Price (EUR)
126,65
Online exclusive
141,00
Save 14,35 (10%)
Each
Quantity:
500 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (EUR)
126,65
Online exclusive
141,00
Save 14,35 (10%)
Each
Chemical Identifiers
CAS156-54-7
IUPAC Namesodium butanoate
Molecular FormulaC4H7NaO2
InChI KeyMFBOGIVSZKQAPD-UHFFFAOYSA-M
SMILES[Na+].CCCC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with HClO497.5 to 102.5 %
Appearance (Form)Powder
Appearance (Color)White
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only