Phenylethyl isothiocyanate, 96%
Phenylethyl isothiocyanate, 96%
Phenylethyl isothiocyanate, 96%
Thermo Scientific Chemicals

Phenylethyl isothiocyanate, 96%

CAS: 2257-09-2 | C9H9NS | 163.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 265160250
Price (EUR)
124,00
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
124,00
Each
Chemical Identifiers
CAS2257-09-2
IUPAC Name(2-isothiocyanatoethyl)benzene
Molecular FormulaC9H9NS
InChI KeyIZJDOKYDEWTZSO-UHFFFAOYSA-N
SMILESS=C=NCCC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Refractive index1.5860 to 1.5910 (20°C, 589 nm)
Appearance (Form)Liquid
GC>=95 %
Appearance (Color)Clear colorless to yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only