2,2,4-Trimethylpentane, 99.5%, for analysis
2,2,4-Trimethylpentane, 99.5%, for analysis
2,2,4-Trimethylpentane, 99.5%, for analysis
Thermo Scientific Chemicals

2,2,4-Trimethylpentane, 99.5%, for analysis

CAS: 540-84-1 | C8H18 | 114.232 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
25 L
Packaging:
View all
Glass Bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 265440025
Price (EUR)
-
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS540-84-1
IUPAC Name2,2,4-trimethylpentane
Molecular FormulaC8H18
InChI KeyNHTMVDHEPJAVLT-UHFFFAOYSA-N
SMILESCC(C)CC(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Cadmium (Cd)=<0.1 ppm
Lead (Pb)=<0.1 ppm
Zinc (Zn)=<0.1 ppm
H2S+foreign org. S cmpds=<50 ppm
Appearance (Form)Clear liquid
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only