Albumine bovine serum, for biochemistry, additional reagent for IGSS, protease free
Albumine bovine serum, for biochemistry, additional reagent for IGSS, protease free
Thermo Scientific Chemicals

Albumine bovine serum, for biochemistry, additional reagent for IGSS, protease free

Albumin, bovine serum (BSA), CAS # 9048-46-8, is a small, stable biomolecule derived from bovine blood with wide applications in biochemistry, immunochemistry, pharmacokinetics, and cell culture studies. | CAS: 9048-46-8
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
100 g
500 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 268131000
Price (EUR)
-
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS9048-46-8
MDL NumberMFCD00145743
SpecificationsSpecification SheetSpecification Sheet
pH6.7 to 7.3 (10 % solution)
Appearance (Form)Lyophilized powder
Loss on drying=<5.0 %
Sodium (Na)=<50 meq/l (10 % solution)
IgGNone detected
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General description

  • Albumin, bovine serum (BSA) is available as a protease-free, lyophilized powder that may be stably stored at 2-8°c
  • It is comprised of 583 amino acid residues, and has a molecular weight of 66.5 kDa

Applications

  • BSA is used as a protein standard in quantification assays, such as the Bradford protein assay
  • It is used to stabilize enzymes in restriction digestions in molecular biology experiments
  • It is a blocking agent in immunoblots thus increasing signal to noise ratios by quenching non-specific immunochemical reactions
  • BSA is a nutritive agent and used as a supplement in cell and microbial culture media
RUO – Research Use Only