Triphenylphosphonium bromide, 99%
Triphenylphosphonium bromide, 99%
Triphenylphosphonium bromide, 99%
Thermo Scientific Chemicals

Triphenylphosphonium bromide, 99%

CAS: 6399-81-1 | C18H16BrP | 343.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 269090250
Price (EUR)
75,00
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
75,00
Each
Chemical Identifiers
CAS6399-81-1
IUPAC Nametriphenylphosphane hydrobromide
Molecular FormulaC18H16BrP
InChI KeyCMSYDJVRTHCWFP-UHFFFAOYSA-N
SMILESBr.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)White to light beige
Appearance (Form)Crystalline powder and/or chunks
Titration Argentometric>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For C-C Bond Formation (Olefination)
RUO – Research Use Only