Potassium tetrafluoroborate, 99%, extra pure
Potassium tetrafluoroborate, 99%, extra pure
Potassium tetrafluoroborate, 99%, extra pure
Thermo Scientific Chemicals

Potassium tetrafluoroborate, 99%, extra pure

CAS: 14075-53-7 | BF4K | 125.90 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 kg
5 kg
Packaging:
View all
Plastic bottle
Catalog number 269220010
Price (EUR)
115,00
Each
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (EUR)
115,00
Each
Chemical Identifiers
CAS14075-53-7
IUPAC Namepotassium tetrafluoroboranuide
Molecular FormulaBF4K
InChI KeyAKEBROIVCDHVSD-UHFFFAOYSA-N
SMILES[K+].F[B-](F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light grey
Silicate (as SiO2)=<0.05 %
Appearance (Form)Crystalline powder
Heavy metals (as Pb)=<30 ppm
Chloride (Cl)=<1000 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent
RUO – Research Use Only