Sodium phosphate, dibasic dodecahydrate, 98.5%, for analysis
Sodium phosphate, dibasic dodecahydrate, 98.5%, for analysis
Sodium phosphate, dibasic dodecahydrate, 98.5%, for analysis
Sodium phosphate, dibasic dodecahydrate, 98.5%, for analysis
Thermo Scientific Chemicals

Sodium phosphate, dibasic dodecahydrate, 98.5%, for analysis

CAS: 10039-32-4 | HNa2O412H2O | 358.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
2.5 kg
10 kg
Packaging:
View all
Plastic bottle
Catalog number 271060025
Price (EUR)
99,10
Each
Add to cart
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (EUR)
99,10
Each
Add to cart
Chemical Identifiers
CAS10039-32-4
IUPAC Namedisodium dodecahydrate hydrogen phosphate
Molecular FormulaH25Na2O16P
InChI KeyDGLRDKLJZLEJCY-UHFFFAOYSA-L
SMILESO.O.O.O.O.O.O.O.O.O.O.O.[Na+].[Na+].OP([O-])([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Heavy metals (as Pb)=<5 ppm
Nitrogen compounds (as N)=<0.001 %
Chloride (Cl)=<5 ppm
Cadmium (Cd)=<5 ppm
Iron (Fe)=<5 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only