2,4-Dimethylstyrene, 97%, stabilized
2,4-Dimethylstyrene, 97%, stabilized
2,4-Dimethylstyrene, 97%, stabilized
Thermo Scientific Chemicals

2,4-Dimethylstyrene, 97%, stabilized

CAS: 2234-20-0 | C10H12 | 132.21 g/mol
Have Questions?
Catalog NumberQuantityPackaging
2711500505 gGlass bottle
Catalog number 271150050
Price (EUR)
75,30
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
75,30
Each
Chemical Identifiers
CAS2234-20-0
IUPAC Name1-ethenyl-2,4-dimethylbenzene
Molecular FormulaC10H12
InChI KeyOEVVKKAVYQFQNV-UHFFFAOYSA-N
SMILESCC1=CC=C(C=C)C(C)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)Clear colorless to yellow
GC>=96.0 %
Stabilizerca. 0.05 % t-Butylcatechol
Appearance (Form)Liquid
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only