5-Hydroxyisophthalic acid, 99%
5-Hydroxyisophthalic acid, 99%
5-Hydroxyisophthalic acid, 99%
Thermo Scientific Chemicals

5-Hydroxyisophthalic acid, 99%

CAS: 618-83-7 | C8H6O5 | 182.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 271391000
Price (EUR)
73,50
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
73,50
Each
Chemical Identifiers
CAS618-83-7
IUPAC Name5-hydroxybenzene-1,3-dicarboxylic acid
Molecular FormulaC8H6O5
InChI KeyQNVNLUSHGRBCLO-UHFFFAOYSA-N
SMILESOC(=O)C1=CC(=CC(O)=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Appearance (Form)Powder
Infrared spectrumConforms
Melting point297°C to 303°C
Titration with NaOH>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only