BOC-L-Tyrosine, 99+%
BOC-L-Tyrosine, 99+%
BOC-L-Tyrosine, 99+%
Thermo Scientific Chemicals

BOC-L-Tyrosine, 99+%

CAS: 3978-80-1 | C14H19NO5 | 281.31 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass Bottle
Catalog number 275700250
Price (EUR)
140,00
Each
Quantity:
25 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (EUR)
140,00
Each
Chemical Identifiers
CAS3978-80-1
IUPAC Name(2S)-2-{[(tert-butoxy)carbonyl]amino}-3-(4-hydroxyphenyl)propanoic acid
Molecular FormulaC14H19NO5
InChI KeyCNBUSIJNWNXLQQ-LDGXTIHJNA-N
SMILESCC(C)(C)OC(=O)N[C@@H](CC1=CC=C(O)C=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
HPLC>=99.0 %
Water=<1.0 % (K.F.)
Infrared spectrumConforms
Specific optical rotation+36.5° to +39.5° (24°C, 589 nm) (c=1, dioxane)
Appearance (Form)Powder
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only