Sodium bis(trimethylsilyl)amide, pure, 2M solution in THF, AcroSeal™
Sodium bis(trimethylsilyl)amide, pure, 2M solution in THF, AcroSeal™
Sodium bis(trimethylsilyl)amide, pure, 2M solution in THF, AcroSeal™
Thermo Scientific Chemicals

Sodium bis(trimethylsilyl)amide, pure, 2M solution in THF, AcroSeal™

CAS: 1070-89-9 | C6H18NNaSi2 | 183.377 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
800 mL
Packaging:
View all
AcroSeal™ Glass bottle
Catalog number 277851000
Price (EUR)
101,00
Each
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
Request bulk or custom format
Price (EUR)
101,00
Each
Chemical Identifiers
CAS1070-89-9
IUPAC Namesodium bis(trimethylsilyl)azanide
Molecular FormulaC6H18NNaSi2
InChI KeyWRIKHQLVHPKCJU-UHFFFAOYSA-N
SMILES[Na+].C[Si](C)(C)[N-][Si](C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with HClO438 to 42 %
Appearance (Color)Clear orange to brown
Appearance (Form)Solution
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only