1,2-Dimethyl-3-hydroxy-4-pyridone, 98+%
1,2-Dimethyl-3-hydroxy-4-pyridone, 98+%
1,2-Dimethyl-3-hydroxy-4-pyridone, 98+%
Thermo Scientific Chemicals

1,2-Dimethyl-3-hydroxy-4-pyridone, 98+%

CAS: 30652-11-0 | C7H9NO2 | 139.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
10 g
50 g
Packaging:
Glass bottle
Catalog number 278740500
Price (EUR)
418,00
Each
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
418,00
Each
Chemical Identifiers
CAS30652-11-0
IUPAC Name3-hydroxy-1,2-dimethyl-1,4-dihydropyridin-4-one
Molecular FormulaC7H9NO2
InChI KeyTZXKOCQBRNJULO-UHFFFAOYSA-N
SMILESCN1C=CC(=O)C(O)=C1C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to almost white
Appearance (Form)Crystalline powder or powder or needles
Infrared spectrumConforms
Titration with HClO498.0 to 102.0 %
HPLC>=98.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only