Potassium sorbate, 99%
Potassium sorbate, 99%
Potassium sorbate, 99%
Thermo Scientific Chemicals

Potassium sorbate, 99%

CAS: 24634-61-5 | C6H7KO2 | 150.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
250 g
1 kg
5 kg
Packaging:
View all
Plastic bottle
Catalog number 291742500
Price (EUR)
32,65
Online exclusive
36,60
Save 3,95 (11%)
Each
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (EUR)
32,65
Online exclusive
36,60
Save 3,95 (11%)
Each
Chemical Identifiers
CAS24634-61-5
IUPAC Namepotassium (2E,4E)-hexa-2,4-dienoate
Molecular FormulaC6H7KO2
InChI KeyCHHHXKFHOYLYRE-STWYSWDKSA-M
SMILES[K+].C\C=C\C=C\C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
Appearance (Form)Powder or granules
Infrared spectrumConforms
Titration with HClO498.5 to 101.5 %
Loss on drying=<1 % (105°C, 3 h)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only