2-Adamantanone, 98%
2-Adamantanone, 98%
2-Adamantanone, 98%
Thermo Scientific Chemicals

2-Adamantanone, 98%

CAS: 700-58-3 | C10H14O | 150.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 292501000
Price (EUR)
281,00
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
281,00
Each
Chemical Identifiers
CAS700-58-3
IUPAC Nameadamantan-2-one
Molecular FormulaC10H14O
InChI KeyIYKFYARMMIESOX-UHFFFAOYSA-N
SMILESO=C1C2CC3CC(C2)CC1C3
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Infrared spectrumConforms
Appearance (Form)Crystalline powder
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only