1,4,7-Triazacyclononane trihydrochloride, 97%
1,4,7-Triazacyclononane trihydrochloride, 97%
1,4,7-Triazacyclononane trihydrochloride, 97%
Thermo Scientific Chemicals

1,4,7-Triazacyclononane trihydrochloride, 97%

CAS: 58966-93-1 | C6H18N3 | 132.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mg
2.5 g
Packaging:
Glass bottle
Catalog number 294310025
Price (EUR)
396,00
Each
Quantity:
2.5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
396,00
Each
Chemical Identifiers
CAS58966-93-1
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Titration Argentometric>=96.0 %
Appearance (Color)Off-white to yellow
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chelation/Complexation reagent
RUO – Research Use Only