3,4-Dichlorobenzaldehyde, 98%
3,4-Dichlorobenzaldehyde, 98%
3,4-Dichlorobenzaldehyde, 98%
Thermo Scientific Chemicals

3,4-Dichlorobenzaldehyde, 98%

CAS: 6287-38-3 | C7H4Cl2O | 175.01 g/mol
Have Questions?
Catalog NumberQuantityPackaging
29456025025 gGlass bottle
Catalog number 294560250
Price (EUR)
54,00
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
54,00
Each
Chemical Identifiers
CAS6287-38-3
IUPAC Name3,4-dichlorobenzaldehyde
Molecular FormulaC7H4Cl2O
InChI KeyZWUSBSHBFFPRNE-UHFFFAOYSA-N
SMILESClC1=CC=C(C=O)C=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Melting point40°C to 44°C
Appearance (Form)Low melting solid
Infrared spectrumConforms
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only