Boron tribromide, 99.9%
Boron tribromide, 99.9%
Boron tribromide, 99.9%
Thermo Scientific Chemicals

Boron tribromide, 99.9%

CAS: 10294-33-4 | BBr3 | 250.52 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 295200250
Price (EUR)
73,65
Online exclusive
82,10
Save 8,45 (10%)
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
73,65
Online exclusive
82,10
Save 8,45 (10%)
Each
Chemical Identifiers
CAS10294-33-4
IUPAC Nametribromoborane
Molecular FormulaBBr3
InChI KeyILAHWRKJUDSMFH-UHFFFAOYSA-N
SMILESBrB(Br)Br
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to amber
Appearance (Form)Liquid
Assay>=99.85 % (By difference)
Heavy metals (as Pb)=<100 ppm
Sulfur (S)=<100 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent, Lewis acid
RUO – Research Use Only