Methyl sulfoxide, 99.8+%, extra pure, Thermo Scientific Chemicals
Methyl sulfoxide, 99.8+%, extra pure, Thermo Scientific Chemicals
Methyl sulfoxide, 99.8+%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methyl sulfoxide, 99.8+%, extra pure, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityPackaging
295522500250mLGlass bottle
2955200101LGlass bottle
2955200252.5LGlass bottle
29552025025LMetal Drum
Catalog number 295522500
Price (EUR)
29,65
Online Exclusive
33,00
Save 3,35 (10%)
Each
Quantity:
250mL
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
29,65
Online Exclusive
33,00
Save 3,35 (10%)
Each
Chemical Identifiers
CAS67-68-5
IUPAC Namemethanesulfinylmethane
Molecular FormulaC2H6OS
InChI KeyIAZDPXIOMUYVGZ-UHFFFAOYSA-N
SMILESCS(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Color scale=<10 APHA
Water=<0.1 %
Acidity=<0.04 mg KOH/g
Appearance (Form)Clear liquid
GC>=99.80 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only