4-tert-Butyltoluene, 96%
4-tert-Butyltoluene, 96%
4-tert-Butyltoluene, 96%
Thermo Scientific Chemicals

4-tert-Butyltoluene, 96%

CAS: 98-51-1 | C11H16 | 148.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
1 L
2.5 L
Packaging:
Glass bottle
Catalog number 296320025
Price (EUR)
195,00
Each
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
195,00
Each
Chemical Identifiers
CAS98-51-1
IUPAC Name1-tert-butyl-4-methylbenzene
Molecular FormulaC11H16
InChI KeyQCWXDVFBZVHKLV-UHFFFAOYSA-N
SMILESCC1=CC=C(C=C1)C(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Refractive index1.4910 to 1.4930 (20°C, 589 nm)
Appearance (Form)Liquid
GC>=95 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only